Assay Design
Gene Transcripts
- Gene Symbol
- CDC42SE1
- Entrez Gene ID
-
56882
- Gene Name
- CDC42 small effector 1
- Gene Aliases
-
SCIP1, SPEC1
- Chromosome Location
-
Chr.1: 151050971 - 151059649 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK054914.1 |
- |
1 - 1 |
1572 |
153 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AI275994.1 |
- |
1 |
|
|
Chr. 1 - 151057758 - 151058235 [+] on Build GRCh38 |
153 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
chromosome 1 open reading frame 56 |
C1orf56 |
MENT |
-6.3kb
|
Sense |
Chr. - 151047783 - 151051420 on Build GRCh38 |
Downstream |
myeloid/lymphoid or mixed-lineage leukemia; translocated to, 11 |
MLLT11 |
AF1Q |
1.4kb
|
Sense |
Chr. - 151059675 - 151068497 on Build GRCh38 |
View Full Product Details