Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_206810.3 | NP_996533.2 | 4 - 5 | 806 | 88 |
NM_206812.3 | NP_996535.2 | 4 - 5 | 806 | 88 | |
GenBank mRNA | AY566846.1 | - | 3 - 4 | 223 | 88 |
U18799.1 | - | 4 - 5 | 577 | 88 | |
U18803.1 | - | 4 - 5 | 577 | 88 | |
U64567.1 | - | 3 - 4 | 490 | 88 | |
U64568.1 | - | 3 - 4 | 490 | 88 |