Assay Design



Gene Transcripts

Gene Symbol
MOG
Entrez Gene ID
4340
Gene Name
myelin oligodendrocyte glycoprotein
Gene Aliases
BTN6, BTNL11, MOGIG2, NRCLP7
Chromosome Location
Chr.6: 29656981 - 29672372 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008228.2 NP_001008229.1 5 - 6 819 111
NM_001170418.1 NP_001163889.1 4 - 5 471 111
NM_206811.3 NP_996534.2 5 - 6 819 111
GenBank mRNA AY566847.1 - 4 - 5 236 111
AY566854.1 - 5 - 6 584 111
U18840.1 - 5 - 6 590 111
U18843.1 - 5 - 6 590 111
U64566.1 - 4 - 5 503 111

View Full Product Details