Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000544.3 | NP_000535.3 | 6 - 7 | 1269 | 70 |
NM_001290043.1 | NP_001276972.1 | 6 - 7 | 1322 | 70 | |
NM_018833.2 | NP_061313.2 | 6 - 7 | 1269 | 70 | |
GenBank mRNA | AB073779.1 | - | 5 - 6 | 1147 | 70 |
AB208953.1 | - | 6 - 7 | 1216 | 70 | |
AF078671.1 | - | 6 - 7 | 1226 | 70 | |
AF105151.1 | - | 6 - 7 | 70 | ||
AF176984.1 | - | 1 - 2 | 202 | 70 | |
AK222823.1 | - | 5 - 6 | 70 | ||
AK223300.1 | - | 6 - 7 | 70 | ||
AK225657.1 | - | 6 - 7 | 70 | ||
AK225693.1 | - | 6 - 7 | 70 | ||
AK292963.1 | - | 6 - 7 | 1252 | 70 | |
BC002751.2 | - | 5 - 6 | 1155 | 70 | |
BT009906.1 | - | 5 - 6 | 1147 | 70 | |
FM246456.1 | - | 5 - 6 | 1147 | 70 | |
M74447.1 | - | 6 - 7 | 70 | ||
M84748.1 | - | 6 - 7 | 1175 | 70 | |
U07844.1 | - | 5 - 6 | 70 | ||
Z22935.1 | - | 6 - 7 | 1175 | 70 | |
Z22936.1 | - | 6 - 7 | 70 |