Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012173.3 | NP_036305.2 | 7 - 8 | 1057 | 121 |
NM_183420.1 | NP_904356.1 | 8 - 9 | 1107 | 121 | |
NM_183421.1 | NP_904357.1 | 8 - 9 | 1107 | 121 | |
XM_011534748.2 | XP_011533050.1 | 8 - 9 | 880 | 121 | |
XM_011534749.1 | XP_011533051.1 | 7 - 8 | 817 | 121 | |
XM_017013313.1 | XP_016868802.1 | 8 - 9 | 931 | 121 | |
XM_017013314.1 | XP_016868803.1 | 7 - 8 | 1009 | 121 | |
GenBank mRNA | AB070270.1 | - | 7 - 8 | 841 | 121 |
AF174605.1 | - | 4 - 5 | 583 | 121 | |
AF419858.1 | - | 8 - 9 | 1107 | 121 | |
AK302334.1 | - | 9 - 10 | 782 | 121 | |
BC020249.2 | - | 6 - 7 | 799 | 121 | |
BC050393.1 | - | 8 - 9 | 960 | 121 |