Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007527.1 | NP_001007528.1 | 13 - 14 | 2062 | 70 |
XM_011514162.2 | XP_011512464.1 | 13 - 14 | 2504 | 70 | |
XM_017010024.1 | XP_016865513.1 | 14 - 15 | 2180 | 70 | |
GenBank mRNA | AK091295.1 | - | 13 - 14 | 2002 | 70 |
AK300433.1 | - | 14 - 15 | 1972 | 70 | |
AK307575.1 | - | 13 - 14 | 2008 | 70 | |
AL137370.1 | - | 2 - 3 | 178 | 70 | |
BC110506.2 | - | 13 - 14 | 2001 | 70 | |
BX647340.1 | - | 13 - 14 | 2037 | 70 | |
CR749399.1 | - | 13 - 14 | 2061 | 70 |