Assay Design



Gene Transcripts

Gene Symbol
TTN
Entrez Gene ID
7273
Gene Name
titin
Gene Aliases
CMD1G, CMH9, CMPD4, EOMFC, HMERF, LGMD2J, MYLK5, TMD
Chromosome Location
Chr.2: 178525989 - 178807423 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256850.1 NP_001243779.1 10 - 11 1894 117
NM_001267550.2 NP_001254479.2 10 - 11 1894 117
NM_133378.4 NP_596869.4 10 - 11 1894 117
NM_133379.4 NP_596870.2 10 - 11 1894 117
XM_017004819.1 XP_016860308.1 10 - 11 1897 117
XM_017004820.1 XP_016860309.1 10 - 11 1897 117
XM_017004821.1 XP_016860310.1 10 - 11 1894 117
XM_017004822.1 XP_016860311.1 10 - 11 1897 117
XM_017004823.1 XP_016860312.1 10 - 11 1897 117
GenBank mRNA X98115.1 - 4 - 5 457 117

View Full Product Details