Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004540.3 | NP_004531.2 | 15 - 16 | 2323 | 67 |
XM_011529575.2 | XP_011527877.1 | 16 - 17 | 2798 | 67 | |
XM_011529576.2 | XP_011527878.1 | 16 - 17 | 2375 | 67 | |
XM_011529579.2 | XP_011527881.1 | 15 - 16 | 2299 | 67 | |
XM_011529585.2 | XP_011527887.1 | 12 - 13 | 1665 | 67 | |
XM_017028353.1 | XP_016883842.1 | 16 - 17 | 2463 | 67 | |
XM_017028356.1 | XP_016883845.1 | 15 - 16 | 2198 | 67 | |
XM_017028357.1 | XP_016883846.1 | 15 - 16 | 2198 | 67 | |
GenBank mRNA | AK299054.1 | - | 12 - 13 | 1836 | 67 |
BC052946.1 | - | 15 - 16 | 2323 | 67 | |
U75330.1 | - | 15 - 16 | 2143 | 67 |