Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004419.4 | NP_001004419.1 | 4 - 5 | 480 | 79 |
GenBank mRNA | AF285089.1 | - | 3 - 4 | 390 | 79 |
AK310150.1 | - | 4 - 5 | 551 | 79 | |
DA814760.1 | - | 4 - 5 | 475 | 79 | |
DB111977.1 | - | 4 - 5 | 481 | 79 | |
KF971857.1 | - | 4 - 5 | 453 | 79 |