Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003061.2 | NP_003052.2 | 6 - 7 | 807 | 89 |
GenBank mRNA | AB011537.3 | - | 6 - 7 | 807 | 89 |
AB017167.1 | - | 6 - 7 | 787 | 89 | |
AY029183.1 | - | 6 - 7 | 504 | 89 | |
BC146761.1 | - | 6 - 7 | 807 | 89 | |
BC146851.1 | - | 6 - 7 | 720 | 89 |