Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003664.4 | NP_003655.3 | 1 - 2 | 303 | 98 |
XM_005248618.3 | XP_005248675.1 | 1 - 2 | 275 | 98 | |
XM_005248619.4 | XP_005248676.1 | 1 - 2 | 275 | 98 | |
GenBank mRNA | AK225908.1 | - | 1 - 2 | 179 | 98 |
AK291201.1 | - | 1 - 2 | 244 | 98 | |
AL708147.1 | - | 1 - 2 | 265 | 98 | |
AY623424.1 | - | 1 - 2 | 205 | 98 | |
BC038444.1 | - | 1 - 2 | 264 | 98 | |
DA889915.1 | - | 1 - 2 | 303 | 98 | |
KU178426.1 | - | 1 - 2 | 127 | 98 | |
KU178427.1 | - | 1 - 2 | 127 | 98 | |
KU178428.1 | - | 1 - 2 | 127 | 98 | |
U81504.1 | - | 1 - 2 | 219 | 98 | |
U91931.1 | - | 1 - 2 | 180 | 98 |