Assay Design



Gene Transcripts

Gene Symbol
CCDC81
Entrez Gene ID
60494
Gene Name
coiled-coil domain containing 81
Gene Aliases
-
Chromosome Location
Chr.11: 86374736 - 86423109 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001156474.1 NP_001149946.1 12 - 13 1895 100
NM_021827.4 NP_068599.3 11 - 12 1625 100
XM_005274161.1 XP_005274218.1 11 - 12 1751 100
XM_005274162.3 XP_005274219.1 7 - 8 796 100
XM_017018105.1 XP_016873594.1 10 - 11 1249 100
GenBank mRNA AK027167.1 - 7 - 8 923 100
AK131331.1 - 11 - 12 1628 100
AK223168.1 - 7 - 8 929 100
AK298875.1 - 8 - 9 944 100
BC126412.1 - 12 - 13 1739 100

View Full Product Details