Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003013.2 | NP_003004.1 | 2 - 3 | 827 | 106 |
GenBank mRNA | AF311912.1 | - | 2 - 3 | 823 | 106 |
AK075372.1 | - | 2 - 3 | 820 | 106 | |
AK093922.1 | - | 3 - 4 | 795 | 106 | |
AY359001.1 | - | 2 - 3 | 629 | 106 | |
BC008666.1 | - | 2 - 3 | 807 | 106 |