Assay Design
Gene Transcripts
- Gene Symbol
- IL17RE
- Entrez Gene ID
-
132014
- Gene Name
- interleukin 17 receptor E
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 9902086 - 9916402 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_153481.1 |
NP_705614.1 |
1 - 2 |
240 |
130 |
GenBank mRNA |
AK096801.1 |
- |
1 - 2 |
220 |
130 |
View Full Product Details