Assay Design



Gene Transcripts

Gene Symbol
FASTKD2
Entrez Gene ID
22868
Gene Name
FAST kinase domains 2
Gene Aliases
KIAA0971
Chromosome Location
Chr.2: 206765388 - 206868284 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014929.3 NP_055744.2 1 - 2 262 96
GenBank mRNA AK001516.1 - 1 - 2 42 96
AL833877.1 - 1 - 2 36 96
DA475994.1 - 1 - 2 293 96

View Full Product Details