Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017759.4 | NP_060229.3 | 6 - 7 | 1705 | 100 |
XM_011511369.2 | XP_011509671.1 | 6 - 7 | 1698 | 100 | |
XM_011511370.2 | XP_011509672.1 | 6 - 7 | 1610 | 100 | |
XM_011511371.2 | XP_011509673.1 | 6 - 7 | 1537 | 100 | |
XM_011511373.2 | XP_011509675.1 | 6 - 7 | 1539 | 100 | |
XM_011511375.2 | XP_011509677.1 | 6 - 7 | 1423 | 100 | |
XM_011511376.2 | XP_011509678.1 | 6 - 7 | 1410 | 100 | |
GenBank mRNA | AK000316.1 | - | 3 - 4 | 1026 | 100 |
AK096585.1 | - | 6 - 7 | 1706 | 100 | |
AK225339.1 | - | 3 - 4 | 1026 | 100 | |
BC004193.2 | - | 5 - 6 | 1422 | 100 | |
BC011687.2 | - | 5 - 6 | 1422 | 100 | |
BC017290.2 | - | 5 - 6 | 1422 | 100 | |
BC136278.1 | - | 5 - 6 | 1403 | 100 |