Assay Design



Gene Transcripts

Gene Symbol
ANKMY1
Entrez Gene ID
51281
Gene Name
ankyrin repeat and MYND domain containing 1
Gene Aliases
ZMYND13
Chromosome Location
Chr.2: 240468632 - 240561110 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282771.1 NP_001269700.1 10 - 11 2303 78
NM_001282780.1 NP_001269709.1 8 - 9 1613 78
NM_001282781.1 NP_001269710.1 9 - 10 1319 78
NM_001308375.1 NP_001295304.1 8 - 9 1852 78
NM_016552.3 NP_057636.2 9 - 10 2140 78
XM_011511300.2 XP_011509602.1 10 - 11 2487 78
XM_011511301.1 XP_011509603.1 10 - 11 2487 78
XM_011511305.2 XP_011509607.1 10 - 11 2487 78
XM_011511309.2 XP_011509611.1 10 - 11 2487 78
XM_011511313.2 XP_011509615.1 10 - 11 2487 78
XM_011511315.2 XP_011509617.1 10 - 11 2487 78
XM_011511317.1 XP_011509619.1 8 - 9 1797 78
XM_011511322.1 XP_011509624.1 8 - 9 1797 78
XM_017004271.1 XP_016859760.1 10 - 11 2487 78
XM_017004272.1 XP_016859761.1 10 - 11 2487 78
XM_017004273.1 XP_016859762.1 10 - 11 2487 78
XM_017004275.1 XP_016859764.1 10 - 11 2253 78
XM_017004276.1 XP_016859765.1 11 - 12 2574 78
XM_017004277.1 XP_016859766.1 10 - 11 2487 78
XM_017004281.1 XP_016859770.1 9 - 10 2093 78
XM_017004282.1 XP_016859771.1 9 - 10 2337 78
XM_017004283.1 XP_016859772.1 11 - 12 2574 78
XM_017004284.1 XP_016859773.1 8 - 9 1797 78
XM_017004285.1 XP_016859774.1 8 - 9 1563 78
XM_017004288.1 XP_016859777.1 7 - 8 1647 78
XM_017004290.1 XP_016859779.1 8 - 9 1864 78
GenBank mRNA AB034636.1 - 9 - 10 2070 78
AK314534.1 - 10 - 11 2287 78
AL833866.1 - 6 - 7 1220 78
BC073146.1 - 8 - 9 1554 78
CR936759.1 - 8 - 9 1521 78

View Full Product Details