Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000492.3 | NP_000483.3 | 22 - 23 | 3856 | 143 |
XM_011515751.2 | XP_011514053.1 | 23 - 24 | 3829 | 143 | |
XM_011515753.1 | XP_011514055.1 | 23 - 24 | 3946 | 143 | |
XM_011515754.2 | XP_011514056.1 | 25 - 26 | 4921 | 143 | |
XM_017011699.1 | XP_016867188.1 | 24 - 25 | 4667 | 143 | |
GenBank mRNA | M28668.1 | - | 22 - 23 | 3856 | 143 |