Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170464.2 | NP_001163935.1 | 3 - 4 | 908 | 85 |
NM_002595.4 | NP_002586.2 | 3 - 4 | 908 | 85 | |
XM_017019405.1 | XP_016874894.1 | 3 - 4 | 350 | 85 | |
XM_017019406.1 | XP_016874895.1 | 3 - 4 | 365 | 85 | |
XM_017019407.1 | XP_016874896.1 | 3 - 4 | 795 | 85 | |
GenBank mRNA | AK290011.1 | - | 3 - 4 | 800 | 85 |
AK294698.1 | - | 2 - 3 | 247 | 85 | |
AK315214.1 | - | 3 - 4 | 818 | 85 | |
BC033005.1 | - | 3 - 4 | 397 | 85 | |
X66360.1 | - | 3 - 4 | 345 | 85 |