Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170464.2 | NP_001163935.1 | 6 - 7 | 1231 | 150 |
NM_002595.4 | NP_002586.2 | 6 - 7 | 1231 | 150 | |
XM_017019405.1 | XP_016874894.1 | 6 - 7 | 673 | 150 | |
XM_017019406.1 | XP_016874895.1 | 6 - 7 | 688 | 150 | |
XM_017019407.1 | XP_016874896.1 | 6 - 7 | 1118 | 150 | |
GenBank mRNA | AK290011.1 | - | 6 - 7 | 1123 | 150 |
AK294698.1 | - | 5 - 6 | 570 | 150 | |
AK315214.1 | - | 6 - 7 | 1141 | 150 | |
BC033005.1 | - | 6 - 7 | 720 | 150 | |
X66360.1 | - | 6 - 7 | 668 | 150 |