Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170464.2 | NP_001163935.1 | 9 - 10 | 1504 | 74 |
NM_002595.4 | NP_002586.2 | 9 - 10 | 1504 | 74 | |
XM_017019405.1 | XP_016874894.1 | 9 - 10 | 946 | 74 | |
XM_017019406.1 | XP_016874895.1 | 9 - 10 | 961 | 74 | |
GenBank mRNA | AK290011.1 | - | 9 - 10 | 1396 | 74 |
AK294698.1 | - | 8 - 9 | 843 | 74 | |
AK315214.1 | - | 9 - 10 | 1414 | 74 | |
BC033005.1 | - | 9 - 10 | 993 | 74 | |
X66360.1 | - | 9 - 10 | 941 | 74 |