Assay Design
Gene Transcripts
- Gene Symbol
- RIC8B
- Entrez Gene ID
-
55188
- Gene Name
- RIC8 guanine nucleotide exchange factor B
- Gene Aliases
-
RIC8, hSyn
- Chromosome Location
-
Chr.12: 106774285 - 106889316 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK001482.1 |
- |
12 - 13 |
1998 |
85 |
AK128102.1 |
- |
11 - 12 |
2538 |
85 |
View Full Product Details