Assay Design



Gene Transcripts

Gene Symbol
RIC8B
Entrez Gene ID
55188
Gene Name
RIC8 guanine nucleotide exchange factor B
Gene Aliases
RIC8, hSyn
Chromosome Location
Chr.12: 106774285 - 106889316 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018157.2 NP_060627.2 4 - 5 966 96
XM_005268998.3 XP_005269055.1 4 - 5 992 96
XM_011538526.2 XP_011536828.1 3 - 4 1348 96
XM_011538527.2 XP_011536829.1 3 - 4 1254 96
XM_011538528.2 XP_011536830.1 3 - 4 1350 96
XM_011538529.2 XP_011536831.1 3 - 4 1345 96
XM_017019548.1 XP_016875037.1 4 - 5 992 96
XM_017019549.1 XP_016875038.1 3 - 4 857 96
GenBank mRNA AK001482.1 - 4 - 5 873 96
AK128102.1 - 2 - 3 1290 96
AY221116.1 - 4 - 5 775 96
BC034220.1 - 5 - 6 1009 96
BC132689.1 - 4 - 5 947 96
BC144056.1 - 4 - 5 947 96
DA118965.1 - 1 - 2 74 96
DA534432.1 - 2 - 3 491 96

View Full Product Details