Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152405.4 | NP_689618.4 | 5 - 6 | 2230 | 99 |
XM_005248430.2 | XP_005248487.1 | 5 - 6 | 2230 | 99 | |
XM_011543155.2 | XP_011541457.1 | 5 - 6 | 2230 | 99 | |
GenBank mRNA | AK095189.1 | - | 4 - 5 | 631 | 99 |
BC130624.1 | - | 4 - 5 | 656 | 99 | |
DB313348.1 | - | 2 - 3 | 99 |