Assay Design



Gene Transcripts

Gene Symbol
SLF1
Entrez Gene ID
84250
Gene Name
SMC5-SMC6 complex localization factor 1
Gene Aliases
ANKRD32, BRCTD1, BRCTx
Chromosome Location
Chr.5: 94613477 - 94697733 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017009980.1 XP_016865469.1 6 - 7 999 233

View Full Product Details