Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128612.2 | NP_001122084.1 | 4 - 5 | 737 | 57 |
NM_001128613.2 | NP_001122085.1 | 3 - 4 | 752 | 57 | |
NM_001271831.1 | NP_001258760.1 | 3 - 4 | 638 | 57 | |
NM_032344.3 | NP_115720.1 | 4 - 5 | 851 | 57 | |
GenBank mRNA | BC006129.1 | - | 4 - 5 | 835 | 57 |
BM923060.1 | - | 3 - 4 | 637 | 57 |