Assay Design
Gene Transcripts
- Gene Symbol
- INTS4
- Entrez Gene ID
-
92105
- Gene Name
- integrator complex subunit 4
- Gene Aliases
-
INT4, MST093
- Chromosome Location
-
Chr.11: 77874491 - 77994849 on Build GRCh38
- Gene Symbol
- INTS4P2
- Entrez Gene ID
-
644619
- Gene Name
- integrator complex subunit 4 pseudogene 2
- Gene Aliases
-
INTS4L2
- Chromosome Location
-
Chr.7: 65647854 - 65718688 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027392.2 |
|
11 - 12 |
1467 |
98 |
- Gene Symbol
- LOC107986705
- Entrez Gene ID
-
107986705
- Gene Name
- integrator complex subunit 4-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 65191623 - 65214524 on Build GRCh38
View Full Product Details