Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001174093.1 | NP_001167564.1 | 1 - 2 | 115 | 139 |
NM_001174096.1 | NP_001167567.1 | 1 - 2 | 115 | 139 | |
NM_001323674.1 | NP_001310603.1 | 1 - 2 | 115 | 139 | |
NM_030751.5 | NP_110378.3 | 1 - 2 | 115 | 139 | |
GenBank mRNA | AK311137.1 | - | 1 - 2 | 76 | 139 |
AK314683.1 | - | 1 - 2 | 76 | 139 | |
BC009572.1 | - | 1 - 2 | 71 | 139 | |
BC107781.1 | - | 1 - 2 | 71 | 139 | |
BC112392.1 | - | 1 - 2 | 68 | 139 | |
DC299346.1 | - | 1 - 2 | 91 | 139 | |
U12170.1 | - | 1 - 2 | 76 | 139 |