Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256738.1 | NP_001243667.1 | 2 - 3 | 298 | 57 |
NM_001256740.1 | NP_001243669.1 | 2 - 3 | 451 | 57 | |
NM_024034.4 | NP_076939.3 | 2 - 3 | 451 | 57 | |
GenBank mRNA | AF123764.1 | - | 2 - 3 | 347 | 57 |
AF123765.1 | - | 2 - 3 | 390 | 57 | |
AF277566.1 | - | 2 - 3 | 406 | 57 | |
AK056251.1 | - | 2 - 3 | 441 | 57 | |
AK225166.1 | - | 2 - 3 | 441 | 57 | |
AK299259.1 | - | 3 - 4 | 363 | 57 | |
BC000199.1 | - | 2 - 3 | 427 | 57 | |
BC009014.1 | - | 2 - 3 | 390 | 57 | |
BP307776.1 | - | 2 - 3 | 451 | 57 | |
BU165271.1 | - | 2 - 3 | 426 | 57 |