Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256737.1 | NP_001243666.1 | 4 - 5 | 779 | 106 |
NM_001256738.1 | NP_001243667.1 | 4 - 5 | 569 | 106 | |
NM_001256739.1 | NP_001243668.1 | 3 - 4 | 548 | 106 | |
NM_024034.4 | NP_076939.3 | 4 - 5 | 722 | 106 | |
NR_046353.1 | 2 - 3 | 355 | 106 | ||
GenBank mRNA | AF123764.1 | - | 4 - 5 | 618 | 103 |
AF123765.1 | - | 4 - 5 | 661 | 103 | |
AF277566.1 | - | 4 - 5 | 677 | 106 | |
AK056251.1 | - | 4 - 5 | 712 | 106 | |
AK225166.1 | - | 4 - 5 | 712 | 106 | |
AK293484.1 | - | 3 - 4 | 498 | 106 | |
AK297348.1 | - | 2 - 3 | 257 | 106 | |
AK299259.1 | - | 5 - 6 | 634 | 106 | |
AK299700.1 | - | 4 - 5 | 728 | 106 | |
BC000199.1 | - | 4 - 5 | 698 | 106 | |
BC009014.1 | - | 4 - 5 | 661 | 106 |