Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145315.1 | NP_001138787.1 | 9 - 10 | 1051 | 75 |
NM_001145316.1 | NP_001138788.1 | 9 - 10 | 1243 | 75 | |
NM_001145317.1 | NP_001138789.1 | 8 - 9 | 730 | 75 | |
NM_001145318.1 | NP_001138790.1 | 8 - 9 | 1002 | 75 | |
NM_024918.3 | NP_079194.3 | 9 - 10 | 1156 | 75 | |
XM_006723876.2 | XP_006723939.1 | 8 - 9 | 725 | 75 | |
XM_017028070.1 | XP_016883559.1 | 9 - 10 | 1003 | 75 | |
XM_017028071.1 | XP_016883560.1 | 9 - 10 | 1225 | 75 | |
XM_017028072.1 | XP_016883561.1 | 8 - 9 | 913 | 75 | |
GenBank mRNA | AK023408.1 | - | 10 - 11 | 881 | 75 |
AK093031.1 | - | 9 - 10 | 1151 | 75 | |
AK290738.1 | - | 9 - 10 | 1243 | 75 | |
AK301671.1 | - | 8 - 9 | 935 | 75 | |
AK301840.1 | - | 8 - 9 | 680 | 75 | |
BC026011.1 | - | 8 - 9 | 1256 | 75 | |
BC058899.1 | - | 9 - 10 | 1009 | 75 |