Assay Design



Gene Transcripts

Gene Symbol
TANGO6
Entrez Gene ID
79613
Gene Name
transport and golgi organization 6 homolog
Gene Aliases
TMCO7
Chromosome Location
Chr.16: 68843547 - 69085182 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024562.1 NP_078838.1 17 - 18 3122 82
GenBank mRNA AB051533.1 - 17 - 18 3122 82
AK022750.1 - 11 - 12 1786 82
AK095333.1 - 16 - 17 2668 82

View Full Product Details