Assay Design



Gene Transcripts

Gene Symbol
TANGO6
Entrez Gene ID
79613
Gene Name
transport and golgi organization 6 homolog
Gene Aliases
TMCO7
Chromosome Location
Chr.16: 68843547 - 69085182 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024562.1 NP_078838.1 4 - 5 1004 83
XM_011523327.2 XP_011521629.1 4 - 5 1063 83
GenBank mRNA AB051533.1 - 4 - 5 1004 83
AK095333.1 - 3 - 4 550 83

View Full Product Details