Assay Design



Gene Transcripts

Gene Symbol
KLHL5
Entrez Gene ID
51088
Gene Name
kelch like family member 5
Gene Aliases
-
Chromosome Location
Chr.4: 39044796 - 39143101 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007075.2 NP_001007076.1 11 - 12 2436 71
NM_001171654.1 NP_001165125.1 10 - 11 1959 71
NM_015990.4 NP_057074.3 10 - 11 2498 71
NM_199039.3 NP_950240.2 9 - 10 2315 71
XM_005262655.4 XP_005262712.1 11 - 12 2470 71
XM_017008275.1 XP_016863764.1 11 - 12 2467 71
GenBank mRNA AB208820.1 - 10 - 11 1731 71
AK001698.1 - 8 - 9 1488 71
AK002174.1 - 9 - 10 1584 71
AK289785.1 - 10 - 11 2498 71
BC048262.1 - 6 - 7 1071 71
BC053860.1 - 11 - 12 2238 71
BC058884.1 - 6 - 7 1001 71
BC081562.1 - 5 - 6 911 71

View Full Product Details