Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021239.2 | NP_067062.1 | 13 - 14 | 1743 | 207 |
XM_011537044.2 | XP_011535346.1 | 14 - 15 | 1845 | 207 | |
XM_011537045.2 | XP_011535347.1 | 13 - 14 | 1655 | 207 | |
GenBank mRNA | AK094697.1 | - | 12 - 13 | 1587 | 207 |
AK125513.1 | - | 4 - 5 | 3208 | 207 | |
AL832314.1 | - | 12 - 13 | 5159 | 207 | |
BC113389.1 | - | 12 - 13 | 1523 | 207 | |
BC113391.1 | - | 12 - 13 | 1523 | 207 | |
BC136775.1 | - | 13 - 14 | 1580 | 207 | |
BC136776.1 | - | 13 - 14 | 1580 | 207 | |
BC144407.1 | - | 14 - 15 | 1648 | 207 | |
BX647116.1 | - | 13 - 14 | 1696 | 207 |