Assay Design



Gene Transcripts

Gene Symbol
BTBD7
Entrez Gene ID
55727
Gene Name
BTB domain containing 7
Gene Aliases
FUP1
Chromosome Location
Chr.14: 93237550 - 93333092 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002860.3 NP_001002860.2 4 - 5 1753 135
NM_001289133.1 NP_001276062.1 2 - 3 485 135
XM_011536939.2 XP_011535241.1 4 - 5 1518 135
GenBank mRNA AB040958.2 - 4 - 5 1682 135
AK021953.1 - 2 - 3 972 135
AK291422.1 - 2 - 3 426 135
BX248762.1 - 2 - 3 432 135

View Full Product Details