Assay Design



Gene Transcripts

Gene Symbol
BTBD7
Entrez Gene ID
55727
Gene Name
BTB domain containing 7
Gene Aliases
FUP1
Chromosome Location
Chr.14: 93237550 - 93333092 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002860.3 NP_001002860.2 10 - 11 2968 142
NM_001289133.1 NP_001276062.1 8 - 9 1700 142
XM_011536939.2 XP_011535241.1 10 - 11 2733 142
XM_017021438.1 XP_016876927.1 7 - 8 1524 142
GenBank mRNA AB040958.2 - 10 - 11 2897 142
AK074354.1 - 1 - 2 443 142
AK291422.1 - 8 - 9 1641 142
AK303355.1 - 6 - 7 1434 142
BX248762.1 - 8 - 9 1647 142
BX538231.1 - 8 - 9 1289 142

View Full Product Details