Assay Design



Gene Transcripts

Gene Symbol
KCTD18
Entrez Gene ID
130535
Gene Name
potassium channel tetramerization domain containing 18
Gene Aliases
6530404F10Rik
Chromosome Location
Chr.2: 200488952 - 200510094 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321550.1 NP_001308479.1 1 - 2 465 123
NM_152387.3 NP_689600.2 1 - 2 465 123
GenBank mRNA BC045189.1 - 1 - 2 436 123
BC059366.1 - 1 - 2 413 123

View Full Product Details