Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321547.1 | NP_001308476.1 | 6 - 7 | 923 | 58 |
NM_001321548.1 | NP_001308477.1 | 6 - 7 | 901 | 58 | |
NM_001321550.1 | NP_001308479.1 | 6 - 7 | 1284 | 58 | |
NM_152387.3 | NP_689600.2 | 6 - 7 | 1306 | 58 | |
GenBank mRNA | AK290867.1 | - | 6 - 7 | 897 | 58 |
BC045189.1 | - | 6 - 7 | 1277 | 58 | |
BC059366.1 | - | 6 - 7 | 1254 | 58 | |
BC067755.1 | - | 6 - 7 | 897 | 58 | |
BU181789.1 | - | 2 - 3 | 162 | 58 |