Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018088.2 | NP_001018098.1 | 49 - 50 | 5953 | 141 |
NM_017684.4 | NP_060154.3 | 47 - 48 | 5824 | 141 | |
NM_018080.3 | NP_060550.2 | 47 - 48 | 5824 | 141 | |
NM_020821.2 | NP_065872.1 | 49 - 50 | 5953 | 141 | |
XM_011521713.2 | XP_011520015.1 | 49 - 50 | 5914 | 141 | |
XM_011521714.2 | XP_011520016.1 | 49 - 50 | 5914 | 141 | |
GenBank mRNA | AJ608770.1 | - | 47 - 48 | 5807 | 141 |
AJ608771.1 | - | 49 - 50 | 5936 | 141 | |
AJ626860.1 | - | 47 - 48 | 5807 | 141 | |
AJ626861.1 | - | 49 - 50 | 5936 | 141 | |
AK128650.1 | - | 3 - 4 | 367 | 141 |