Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291285.1 | NP_001278214.1 | 2 - 3 | 5309 | 85 |
NM_001291303.1 | NP_001278232.1 | 2 - 3 | 5309 | 85 | |
NM_024582.4 | NP_078858.4 | 2 - 3 | 5309 | 85 | |
XM_011532236.2 | XP_011530538.1 | 3 - 4 | 6386 | 85 | |
XM_011532237.2 | XP_011530539.1 | 2 - 3 | 1194 | 85 | |
GenBank mRNA | AY356402.1 | - | 2 - 3 | 203 | 85 |
DT932755.1 | - | 2 - 3 | 661 | 85 |