Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291285.1 | NP_001278214.1 | 9 - 10 | 11800 | 103 |
NM_001291303.1 | NP_001278232.1 | 9 - 10 | 11800 | 103 | |
NM_024582.4 | NP_078858.4 | 9 - 10 | 11794 | 103 | |
XM_011532236.2 | XP_011530538.1 | 10 - 11 | 12877 | 103 | |
XM_011532237.2 | XP_011530539.1 | 9 - 10 | 7685 | 103 | |
GenBank mRNA | AK096646.1 | - | 1 - 2 | 96 | 103 |
AK291461.1 | - | 1 - 2 | 340 | 103 | |
KJ131486.1 | - | 1 - 2 | 37 | 103 |