Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178563.3 | NP_848658.3 | 14 - 15 | 2294 | 118 |
XM_017012136.1 | XP_016867625.1 | 14 - 15 | 3616 | 118 | |
XM_017012137.1 | XP_016867626.1 | 14 - 15 | 3898 | 118 | |
XM_017012138.1 | XP_016867627.1 | 13 - 14 | 2097 | 118 | |
XM_017012139.1 | XP_016867628.1 | 11 - 12 | 1920 | 118 | |
XM_017012141.1 | XP_016867630.1 | 13 - 14 | 3433 | 118 | |
XM_017012142.1 | XP_016867631.1 | 13 - 14 | 2100 | 118 | |
GenBank mRNA | AK302827.1 | - | 14 - 15 | 2294 | 118 |
BC030651.2 | - | 15 - 16 | 2402 | 118 |