Assay Design
Gene Transcripts
- Gene Symbol
- PCDHA13
- Entrez Gene ID
-
56136
- Gene Name
- protocadherin alpha 13
- Gene Aliases
-
CNR5, CNRN5, CNRS5, CRNR5, PCDH-ALPHA13
- Chromosome Location
-
Chr.5: 140882269 - 141012344 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_031865.1 |
NP_114071.1 |
1 - 1 |
2362 |
75 |
GenBank mRNA |
AF152478.1 |
- |
1 - 1 |
2362 |
75 |
View Full Product Details