Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003735.2 | NP_003726.1 | 1 - 1 | 1465 | 106 |
NM_032094.1 | NP_115265.1 | 1 - 1 | 1465 | 106 | |
GenBank mRNA | AB000897.1 | - | 1 - 1 | 366 | 106 |
AB011160.1 | - | 1 - 1 | 1438 | 106 | |
AF152321.1 | - | 1 - 1 | 1296 | 106 | |
AF152506.1 | - | 1 - 1 | 1296 | 106 | |
AY358946.1 | - | 1 - 1 | 1666 | 106 |