Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291661.1 | NP_001278590.1 | 61 - 62 | 10024 | 115 |
XM_011533466.1 | XP_011531768.1 | 61 - 62 | 10009 | 115 | |
XM_011533467.1 | XP_011531769.1 | 61 - 62 | 9976 | 115 | |
XM_011533468.1 | XP_011531770.1 | 61 - 62 | 9946 | 115 | |
XM_011533469.1 | XP_011531771.1 | 60 - 61 | 9868 | 115 | |
XM_017005860.1 | XP_016861349.1 | 61 - 62 | 10009 | 115 | |
XM_017005861.1 | XP_016861350.1 | 60 - 61 | 10692 | 115 | |
XM_017005862.1 | XP_016861351.1 | 40 - 41 | 6584 | 115 | |
GenBank mRNA | AK097746.1 | - | 2 - 3 | 310 | 115 |
AK128592.1 | - | 9 - 10 | 1450 | 115 |