Assay Design



Gene Transcripts

Gene Symbol
NTN3
Entrez Gene ID
4917
Gene Name
netrin 3
Gene Aliases
NTN2L
Chromosome Location
Chr.16: 2471499 - 2474145 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006181.2 NP_006172.1 5 - 6 1601 126
GenBank mRNA U86759.1 - 5 - 6 1601 126

View Full Product Details