Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145873.1 | NP_001139345.1 | 6 - 7 | 1544 | 116 |
NM_001768.6 | NP_001759.3 | 3 - 4 | 1402 | 116 | |
NR_027353.1 | 4 - 5 | 988 | 116 | ||
GenBank mRNA | AK097942.1 | - | 4 - 5 | 587 | 116 |
AK124156.1 | - | 6 - 7 | 1544 | 116 | |
AK300089.1 | - | 4 - 5 | 954 | 116 | |
AY039664.1 | - | 3 - 4 | 692 | 116 | |
BC025715.1 | - | 3 - 4 | 518 | 116 | |
M12824.1 | - | 3 - 4 | 600 | 116 | |
M12828.1 | - | 3 - 4 | 578 | 116 |