Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000095.2 | NP_000086.2 | 18 - 19 | 2261 | 64 |
GenBank mRNA | AB086984.1 | - | 18 - 19 | 2225 | 64 |
AK074508.1 | - | 14 - 15 | 1589 | 64 | |
AK223216.1 | - | 18 - 19 | 2261 | 64 | |
AK290595.1 | - | 18 - 19 | 2261 | 64 | |
AK296586.1 | - | 17 - 18 | 2102 | 64 | |
AK297798.1 | - | 17 - 18 | 2163 | 64 | |
AK297814.1 | - | 17 - 18 | 2222 | 64 | |
BC033676.1 | - | 17 - 18 | 2485 | 64 | |
BC110847.1 | - | 18 - 19 | 2247 | 64 | |
BC125092.1 | - | 18 - 19 | 2254 | 64 | |
L32137.1 | - | 18 - 19 | 2250 | 64 |