Assay Design



Gene Transcripts

Gene Symbol
PCYOX1L
Entrez Gene ID
78991
Gene Name
prenylcysteine oxidase 1 like
Gene Aliases
-
Chromosome Location
Chr.5: 149358007 - 149375112 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301057.1 NP_001287986.1 3 - 4 552 74
GenBank mRNA BC000014.2 - 3 - 4 488 74

View Full Product Details